Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYB SIRNA Assays ›
Gene Symbol:
MYB
Gene Name:
MYB proto-oncogene, transcription factor
Gene Aliases:
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location:
Chr.6: 135180981 - 135219173 on Build GRCh38
UniGene ID:
Hs.606320
Species:
Human
siRNA ID
HSS181383
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130172.1
NP_001123644.1
6
918
NM_001130173.1
NP_001123645.1
6
918
NM_001161656.1
NP_001155128.1
6
918
NM_001161657.1
NP_001155129.1
6
918
NM_001161658.1
NP_001155130.1
6
918
NM_001161659.1
NP_001155131.1
6
918
NM_001161660.1
NP_001155132.1
6
918
NM_005375.3
NP_005366.2
6
918
NR_134958.1
6
918
NR_134959.1
6
918
NR_134960.1
6
918
NR_134961.1
6
918
NR_134962.1
5
697
NR_134963.1
6
918
NR_134964.1
6
918
NR_134965.1
6
918
GenBank mRNA
AF104863.1
-
6
796
AJ606317.1
-
6
918
AJ606318.1
-
6
918
AJ606319.1
-
6
918
AJ606320.1
-
6
918
AJ606321.1
-
6
918
AJ606322.1
-
6
918
AJ606323.1
-
6
918
AJ606324.1
-
6
918
AJ616235.1
-
6
833
AJ616796.4
-
1
192
AY787443.1
-
6
719
AY787444.1
-
4
426
AY787445.1
-
5
647
AY787446.1
-
5
498
AY787447.1
-
6
719
AY787448.1
-
6
719
AY787449.1
-
6
719
AY787450.1
-
6
719
AY787451.1
-
6
719
AY787452.1
-
6
719
AY787453.1
-
6
719
AY787454.1
-
6
719
AY787455.1
-
6
719
AY787456.1
-
6
719
AY787457.1
-
6
719
AY787458.1
-
6
719
AY787459.1
-
6
719
AY787460.1
-
6
719
AY787461.1
-
6
719
AY787462.1
-
6
719
AY787463.1
-
6
719
AY787464.1
-
6
719
AY787465.1
-
6
719
AY787466.1
-
6
719
AY787467.1
-
6
719
AY787468.1
-
6
719
AY787469.1
-
6
719
AY787470.1
-
6
719
AY787471.1
-
6
719
AY787472.1
-
6
719
AY787473.1
-
6
719
AY787474.1
-
6
719
AY787475.1
-
6
719
BC064955.1
-
6
870
6069320
M13665.1
-
4
581
M13666.1
-
4
581
M15024.1
-
6
832
X52125.1
-
6
916
X52126.1
-
5
722
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
chromatin remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of T-helper cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline