Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015675.3 | NP_056490.2 | 2 | 366 | ||
XM_017026822.1 | XP_016882311.1 | 2 | 364 | |||
GenBank mRNA | AF078077.1 | - | 2 | 232 | ||
AF087853.1 | - | 2 | 353 | |||
AF090950.1 | - | 2 | 161 | |||
AK293087.1 | - | 2 | 364 | |||
AY615270.1 | - | 1 | 261 | |||
AY615271.1 | - | 2 | 164 | |||
BC024654.3 | - | 2 | 272 | |||
BC113466.1 | - | 2 | 223 | 8322518 | ||
BC117469.1 | - | 2 | 223 | 40126020 | ||
BP225713.1 | - | 2 | 368 |