Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130065.1 | NP_001123537.1 | 22 | 3829 | ||
NM_004145.3 | NP_004136.2 | 22 | 3829 | |||
GenBank mRNA | AB209997.1 | - | 22 | 3824 | ||
AB290184.1 | - | 22 | 3824 | |||
CN284963.1 | - | 1 | 263 |