Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000262.2 | NP_000253.1 | 8 | 1558 | ||
XM_005261615.4 | XP_005261672.1 | 9 | 1197 | |||
XM_005261616.4 | XP_005261673.1 | 9 | 1179 | |||
GenBank mRNA | BC000095.2 | - | 8 | 1094 | 3504221 | |
CR456527.1 | - | 8 | 1067 | |||
M29276.1 | - | 8 | 1426 | |||
M38083.1 | - | 8 | 1085 | |||
M62783.1 | - | 8 | 1482 |