Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005969.3 | NP_005960.1 | 4, 5 | 310 | ||
GenBank mRNA | AB500094.1 | - | 2, 3 | 169 | ||
AK299515.1 | - | 4, 5 | 233 | |||
AK312599.1 | - | 4, 5 | 318 | |||
AK316242.1 | - | 4, 5 | 227 | |||
AK316324.1 | - | 4, 5 | 254 | |||
AK316548.1 | - | 5, 6 | 367 | |||
BC022090.2 | - | 4, 5 | 220 | 3029455 | ||
U77456.1 | - | 4, 5 | 318 |