Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004545.3 | NP_004536.2 | 3 | 334 | ||
GenBank mRNA | AF050638.1 | - | 3 | 188 | ||
AK055348.1 | - | 3 | 2260 | |||
AK316557.2 | - | 3 | 956 | |||
BC009691.1 | - | 3 | 194 | |||
BC104672.1 | - | 3 | 254 | |||
BC126232.1 | - | 3 | 331 | 8991948 | ||
BC126234.1 | - | 3 | 331 | 8991950 | ||
BM052981.1 | - | 3 | 334 | |||
BX476462.1 | - | 3 | 233 |