Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004546.2 | NP_004537.1 | 3 | 327 | ||
GenBank mRNA | AF050639.1 | - | 3 | 320 | ||
AF067166.1 | - | 3 | 327 | |||
BC001168.1 | - | 3 | 294 | |||
BC063026.1 | - | 4 | 492 | 6064434 | ||
CR542121.1 | - | 3 | 263 |