Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004146.5 | NP_004137.2 | 1 | 149 | ||
GenBank mRNA | AF112200.1 | - | 1 | 135 | ||
AF217091.1 | - | 1 | 94 | |||
BC002595.2 | - | 1 | 134 | 3140806 | ||
BU565399.1 | - | 150 | ||||
CB124371.1 | - | 1 | 149 | |||
CR450354.1 | - | 1 | 72 | |||
M33374.1 | - | 1 | 107 |