Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184986.1 | NP_001171915.1 | 3, 4 | 900 | ||
NM_001184987.1 | NP_001171916.1 | 2, 3 | 368 | |||
NM_001184988.1 | NP_001171917.1 | 3, 4 | 404 | |||
NM_001184989.1 | NP_001171918.1 | 3, 4 | 427 | |||
NM_001184990.1 | NP_001171919.1 | 2, 3 | 364 | |||
NM_001184991.1 | NP_001171920.1 | 2, 3 | 841 | |||
NM_002494.3 | NP_002485.1 | 2, 3 | 503 | |||
GenBank mRNA | AF047184.1 | - | 1, 2 | 98 | ||
AF047435.1 | - | 1, 2 | 78 | |||
AK291147.1 | - | 1, 2 | 108 | |||
BC101798.1 | - | 1, 2 | 114 | 8069304 | ||
BC107682.1 | - | 2, 3 | 473 | 5557684 | ||
BC112061.1 | - | 1, 2 | 114 | 8327529 | ||
BM768825.1 | - | 3, 4 | 404 | |||
BU677336.1 | - | 3, 4 | 316 | |||
CA313899.1 | - | 3, 4 | 353 | |||
CB047240.1 | - | 3, 4 | 363 | |||
CB160001.1 | - | 3, 4 | 355 | |||
CN347371.1 | - | 2, 3 | 269 | |||
CR457005.1 | - | 1, 2 | 70 |