Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004044.6 | NP_004035.2 | 12 | 1333 | ||
XM_017004187.1 | XP_016859676.1 | 12 | 1256 | |||
GenBank mRNA | AB062403.1 | - | 12 | 1181 | ||
AK290067.1 | - | 11 | 1315 | |||
AK298142.1 | - | 12 | 1446 | |||
AK313066.1 | - | 12 | 1227 | |||
BC008879.2 | - | 12 | 1176 | 4300570 | ||
D82348.1 | - | 12 | 1221 | |||
D89976.1 | - | 12 | 1203 | |||
EU794654.1 | - | 12 | 1320 | |||
U37436.1 | - | 11 | 1143 |