Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000702.3 | NP_000693.1 | 21, 22 | 3077 | ||
GenBank mRNA | AB018321.2 | - | 21, 22 | 3034 | ||
AK124581.1 | - | 19, 20 | 2749 | |||
AK126573.1 | - | 14, 15 | 2026 | |||
AK295729.1 | - | 14, 15 | 2054 | |||
AK299460.1 | - | 6, 7 | 828 | |||
AK314296.1 | - | 21, 22 | 3048 | |||
AL831991.1 | - | 20, 21 | 2866 | |||
AL831997.1 | - | 20, 21 | 2892 | |||
BC052271.1 | - | 21, 22 | 3028 | 6672133 | ||
KU177927.1 | - | 20, 21 | 2945 | |||
KU177928.1 | - | 8, 9 | 947 |