Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289999.1 | NP_001276928.1 | 2 | 1212 | ||
NM_001290000.1 | NP_001276929.1 | 2 | 1064 | |||
NM_005384.2 | NP_005375.2 | 2 | 1069 | |||
XM_017014743.1 | XP_016870232.1 | 2 | 1207 | |||
XM_017014744.1 | XP_016870233.1 | 2 | 976 | |||
GenBank mRNA | AK313970.1 | - | 2 | 886 | ||
BC008197.1 | - | 1 | 707 | 3048714 | ||
CR542049.1 | - | 1 | 674 | |||
HM204564.1 | - | 1 | 674 | |||
S79880.1 | - | 2 | 887 | |||
U26173.1 | - | 2 | 887 | |||
X64318.1 | - | 2 | 887 |