Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020529.2 | NP_065390.1 | 5 | 988 | ||
GenBank mRNA | AK311515.1 | - | 4 | 750 | ||
AK313421.1 | - | 5 | 984 | |||
AY033600.1 | - | 5 | 892 | |||
BC002601.2 | - | 5 | 980 | 3161686 | ||
BC004983.1 | - | 5 | 971 | 2957970 | ||
BT007091.1 | - | 5 | 878 | |||
BU737609.1 | - | 2 | 588 | |||
M69043.1 | - | 5 | 972 |