Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF097419.1 | - | 4 | 663 | ||
AF097420.1 | - | 4 | 663 | |||
AF097423.1 | - | 4 | 663 | |||
AF097424.1 | - | 4 | 663 | |||
AF097425.1 | - | 4 | 663 | |||
AF097426.1 | - | 4 | 663 | |||
AF097427.1 | - | 4 | 663 | |||
AF097428.1 | - | 4 | 663 | |||
AF097429.1 | - | 4 | 663 | |||
AK291893.1 | - | 4 | 780 | |||
AK300818.1 | - | 4 | 660 | |||
AK313888.1 | - | 4 | 715 | |||
BC105064.1 | - | 4 | 718 | 8144067 | ||
BC105068.1 | - | 4 | 718 | 8144071 | ||
BC143671.1 | - | 4 | 673 | 9052185 | ||
X77909.1 | - | 4 | 729 |