Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001096.2 | NP_001087.2 | 26, 27 | 3214 | ||
NM_001303274.1 | NP_001290203.1 | 26, 27 | 3221 | |||
NM_001303275.1 | NP_001290204.1 | 25, 26 | 3191 | |||
NM_198830.1 | NP_942127.1 | 25, 26 | 3184 | |||
XM_005257395.1 | XP_005257452.1 | 26, 27 | 3263 | |||
XM_017024688.1 | XP_016880177.1 | 25, 26 | 3233 | |||
GenBank mRNA | AB210035.1 | - | 26, 27 | 3161 | ||
AK095084.1 | - | 16, 17 | 1995 | |||
AK295675.1 | - | 25, 26 | 3191 | |||
AK304802.1 | - | 18, 19 | 2365 | |||
AK312315.1 | - | 26, 27 | 3218 | |||
BC006195.2 | - | 26, 27 | 3182 | 2959339 | ||
U18197.1 | - | 26, 27 | 3122 | |||
X64330.1 | - | 26, 27 | 3147 |