Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006166.3 | NP_006157.1 | 5 | 595 | ||
XM_005268908.4 | XP_005268965.1 | 6 | 659 | |||
XM_005268909.4 | XP_005268966.1 | 6 | 642 | |||
XM_005268910.4 | XP_005268967.1 | 6 | 1314 | |||
GenBank mRNA | AK291934.1 | - | 5 | 527 | ||
BC005316.1 | - | 5 | 544 | 3680937 | ||
BC005317.1 | - | 5 | 550 | 3997299 | ||
BC007035.1 | - | 5 | 522 | 3996244 | ||
L06145.1 | - | 3 | 394 | |||
X59710.1 | - | 5 | 499 |