Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318193.1 | NP_001305122.1 | 2 | 309 | ||
NM_002528.6 | NP_002519.1 | 2 | 309 | |||
XM_017023253.1 | XP_016878742.1 | 2 | 312 | |||
GenBank mRNA | AB001575.1 | - | 2 | 273 | ||
BC000391.2 | - | 2 | 266 | |||
BC003014.1 | - | 2 | 266 | 2821314 | ||
BE900738.1 | - | 2 | 191 | |||
HY057336.1 | - | 2 | 286 | |||
HY094895.1 | - | 2 | 312 | |||
U79718.1 | - | 2 | 306 | |||
U81285.1 | - | 2 | 271 | |||
Y09687.1 | - | 2 | 260 |