Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083592.1 | NP_001077061.1 | 2 | 533 | ||
NM_005012.3 | NP_005003.2 | 2 | 533 | |||
XM_011541526.1 | XP_011539828.1 | 2 | 132 | |||
XM_017001377.1 | XP_016856866.1 | 2 | 2980 | |||
GenBank mRNA | AK308088.1 | - | 2 | 539 | ||
BC080541.1 | - | 2 | 533 | |||
BC128386.1 | - | 3 | 143 | |||
M97675.1 | - | 2 | 509 |