Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001331.2 | NP_001001331.1 | 2 | 564 | ||
NM_001683.3 | NP_001674.2 | 2 | 564 | |||
XM_005265179.4 | XP_005265236.1 | 4 | 748 | |||
XM_006713175.3 | XP_006713238.1 | 4 | 632 | |||
XM_011533752.2 | XP_011532054.1 | 2 | 537 | |||
XM_017006481.1 | XP_016861970.1 | 5 | 875 | |||
XM_017006482.1 | XP_016861971.1 | 2 | 566 | |||
XM_017006483.1 | XP_016861972.1 | 2 | 669 | |||
XM_017006484.1 | XP_016861973.1 | 4 | 632 | |||
XM_017006485.1 | XP_016861974.1 | 2 | 671 | |||
XM_017006486.1 | XP_016861975.1 | 2 | 671 | |||
XM_017006487.1 | XP_016861976.1 | 4 | 631 | |||
XM_017006488.1 | XP_016861977.1 | 2 | 671 | |||
XM_017006489.1 | XP_016861978.1 | 2 | 538 | |||
XM_017006490.1 | XP_016861979.1 | 2 | 553 | |||
XM_017006491.1 | XP_016861980.1 | 2 | 670 | |||
XM_017006492.1 | XP_016861981.1 | 2 | 670 | |||
GenBank mRNA | AB210008.1 | - | 2 | 637 | ||
AK308898.1 | - | 2 | 564 | |||
L00620.1 | - | 1 | 143 | |||
L20977.1 | - | 4 | 701 | |||
M97260.1 | - | 2 | 545 | |||
S95738.1 | - | 1 | 195 | |||
X63575.1 | - | 1 | 195 |