Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243146.1 | NP_001230075.1 | 13 | 1559 | ||
NM_001243147.1 | NP_001230076.1 | 14 | 1610 | |||
NM_002533.3 | NP_002524.2 | 15 | 1883 | |||
NM_206840.2 | NP_996671.1 | 14 | 1809 | |||
XM_011544196.1 | XP_011542498.1 | 16 | 1947 | |||
XM_011544198.1 | XP_011542500.1 | 15 | 1902 | |||
XM_011544199.1 | XP_011542501.1 | 15 | 1856 | |||
XM_011544200.1 | XP_011542502.1 | 14 | 1830 | |||
XM_017001378.1 | XP_016856867.1 | 17 | 2056 | |||
XM_017001379.1 | XP_016856868.1 | 16 | 1975 | |||
XM_017001380.1 | XP_016856869.1 | 17 | 2056 | |||
XM_017001381.1 | XP_016856870.1 | 17 | 2053 | |||
XM_017001382.1 | XP_016856871.1 | 16 | 1986 | |||
XM_017001383.1 | XP_016856872.1 | 15 | 1905 | |||
XM_017001384.1 | XP_016856873.1 | 17 | 2056 | |||
XM_017001385.1 | XP_016856874.1 | 17 | 2056 | |||
XM_017001386.1 | XP_016856875.1 | 13 | 1749 | |||
GenBank mRNA | AK297070.1 | - | 15 | 1842 | ||
AK297396.1 | - | 13 | 1510 | |||
AK298244.1 | - | 14 | 1559 | |||
BC012105.1 | - | 13 | 1470 | 4636004 | ||
U68140.1 | - | 15 | 1877 | |||
U78772.1 | - | 14 | 1790 |