Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008391.3 | NP_001008392.2 | 9 | 687 | ||
XM_011520139.2 | XP_011518441.1 | 8 | 548 | |||
GenBank mRNA | AK131470.1 | - | 9 | 670 | ||
BC062749.1 | - | 9 | 687 |