Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012393.2 | NP_001012393.1 | 7 | 1073 | ||
NM_001319103.1 | NP_001306032.1 | 6 | 1351 | |||
NM_001319104.1 | NP_001306033.1 | 6 | 988 | |||
NM_001319105.1 | NP_001306034.1 | 7 | 897 | |||
NM_001319106.1 | NP_001306035.1 | 6 | 1348 | |||
NM_002545.4 | NP_002536.1 | 6 | 1351 | |||
XM_006718846.2 | XP_006718909.1 | 7 | 1296 | |||
XM_011542856.2 | XP_011541158.1 | 8 | 1609 | |||
GenBank mRNA | AK289695.1 | - | 6 | 979 | ||
AK299528.1 | - | 8 | 746 | |||
AK299908.1 | - | 7 | 860 | |||
AK314077.1 | - | 6 | 1348 | |||
AK315859.1 | - | 6 | 678 | |||
AK316273.1 | - | 6 | 775 | |||
AK316491.1 | - | 7 | 897 | |||
BC074742.2 | - | 6 | 830 | 30915458 | ||
BC074773.2 | - | 6 | 833 | 30915200 | ||
BC117254.1 | - | 6 | 829 | 40125805 | ||
BC126251.1 | - | 6 | 829 | 8991967 | ||
BC143945.1 | - | 6 | 826 | 9052463 | ||
BC143946.1 | - | 6 | 829 | 9052464 | ||
BX537377.1 | - | 7 | 921 | |||
EU562295.1 | - | 6 | 1385 | |||
EU562296.1 | - | 7 | 863 | |||
EU562297.1 | - | 7 | 800 | |||
EU562298.1 | - | 6 | 778 | |||
EU562299.1 | - | 6 | 690 | |||
EU562300.1 | - | 5 | 614 | |||
L34774.1 | - | 6 | 856 |