Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098402.1 | NP_001091872.1 | 3 | 374 | ||
NM_001098403.1 | NP_001091873.1 | 3 | 374 | |||
NM_001320729.1 | NP_001307658.1 | 2 | 309 | |||
NM_001320731.1 | NP_001307660.1 | 4 | 528 | |||
NM_020727.4 | NP_065778.3 | 2 | 309 | |||
XM_005261121.3 | XP_005261178.1 | 3 | 714 | |||
XM_011529588.2 | XP_011527890.1 | 4 | 619 | |||
XM_011529590.2 | XP_011527892.1 | 4 | 928 | |||
XM_017028360.1 | XP_016883849.1 | 3 | 952 | |||
XM_017028361.1 | XP_016883850.1 | 3 | 322 | |||
GenBank mRNA | AB033053.1 | - | 2 | 263 | ||
AB041014.1 | - | 3 | 344 | |||
AB041015.1 | - | 3 | 344 | |||
AK300291.1 | - | 2 | 270 | |||
AK308179.1 | - | 4 | 556 | |||
BC063290.1 | - | 2 | 270 | 30348821 | ||
BP230048.1 | - | 2 | 309 | |||
CR933655.1 | - | 4 | 507 |