Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006190.4 | NP_006181.1 | 17 | 1877 | ||
NR_033915.1 | 16 | 2394 | ||||
XM_006712555.3 | XP_006712618.1 | 17 | 1824 | |||
XM_011511252.2 | XP_011509554.1 | 18 | 1939 | |||
XM_011511253.2 | XP_011509555.1 | 18 | 1811 | |||
GenBank mRNA | BC014834.1 | - | 17 | 1820 | 2899929 | |
BP264347.1 | - | 1 | 396 | |||
U27459.1 | - | 17 | 1811 | |||
U40268.2 | - | 17 | 1792 |