Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315501.1 | NP_001302430.1 | 11 | 1629 | ||
NM_001315502.1 | NP_001302431.1 | 9 | 1119 | |||
NM_002555.5 | NP_002546.3 | 11 | 1413 | |||
NM_183233.2 | NP_899056.2 | 11 | 1416 | |||
GenBank mRNA | AB012083.1 | - | 11 | 1392 | ||
AF028738.1 | - | 11 | 1392 | |||
AF030302.1 | - | 11 | 1629 | |||
AF037064.1 | - | 11 | 1400 | |||
AF059663.1 | - | 11 | 1392 | |||
AF070479.1 | - | 11 | 1392 | |||
AW452749.1 | - | 1 | 139 | |||
BC015571.2 | - | 11 | 1416 | 3636077 | ||
BG684685.1 | - | 3 | 334 |