Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315501.1 | NP_001302430.1 | 2 | 506 | ||
NM_001315502.1 | NP_001302431.1 | 2 | 290 | |||
NM_002555.5 | NP_002546.3 | 2 | 290 | |||
NM_183233.2 | NP_899056.2 | 2 | 293 | |||
XM_011520141.2 | XP_011518443.2 | 2 | 449 | |||
XM_011520142.2 | XP_011518444.2 | 2 | 448 | |||
GenBank mRNA | AB012083.1 | - | 2 | 269 | ||
AF028738.1 | - | 2 | 269 | |||
AF030302.1 | - | 2 | 506 | |||
AF037064.1 | - | 2 | 277 | |||
AF059663.1 | - | 2 | 269 | |||
AF070479.1 | - | 2 | 269 | |||
AL521035.3 | - | 2 | 287 | |||
BC015571.2 | - | 2 | 293 | 3636077 | ||
BG489795.1 | - | 2 | 279 | |||
BM561526.1 | - | 2 | 236 | |||
BP350681.1 | - | 2 | 290 | |||
DA582280.1 | - | 2 | 379 |