Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319108.1 | NP_001306037.1 | 3 | 804 | ||
NM_020530.5 | NP_065391.1 | 3 | 623 | |||
GenBank mRNA | AB451493.1 | - | 2 | 504 | ||
BC011589.1 | - | 3 | 620 | 4548943 | ||
CR456534.1 | - | 3 | 593 | |||
CR541703.1 | - | 3 | 579 | |||
CR981101.1 | - | 3 | 623 |