Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199770.1 | NP_001186699.1 | 5 | 787 | ||
NM_014562.3 | NP_055377.1 | 5 | 787 | |||
NR_130153.1 | 5 | 902 | ||||
GenBank mRNA | AK095680.1 | - | 5 | 759 | ||
AK297308.1 | - | 5 | 873 | |||
BC007621.1 | - | 5 | 731 | 3355563 |