Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002557.3 | NP_002548.3 | 10, 11 | 1215 | ||
GenBank mRNA | AB208783.1 | - | 8, 9 | 1597 | ||
AK295207.1 | - | 7, 8 | 1260 | |||
AK298757.1 | - | 8, 9 | 958 | |||
AK301726.1 | - | 6, 7 | 697 | |||
AK314074.1 | - | 10, 11 | 1173 | |||
AY189737.1 | - | 10, 11 | 1216 | |||
BC126177.1 | - | 10, 11 | 1198 | 8991893 | ||
BC136406.1 | - | 10, 11 | 1198 | 9020393 | ||
U09550.1 | - | 10, 11 | 1171 |