Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002567.3 | NP_002558.1 | 3 | 602 | ||
GenBank mRNA | AK226006.1 | - | 3 | 381 | ||
AK299414.1 | - | 3 | 361 | |||
AK308056.1 | - | 3 | 360 | |||
AK311927.1 | - | 3 | 373 | |||
BC008714.2 | - | 3 | 375 | 2822606 | ||
BC017396.1 | - | 3 | 396 | 4074533 | ||
BC018799.1 | - | 3 | 366 | |||
BC031102.1 | - | 3 | 364 | 5300366 | ||
BF978793.1 | - | 3 | 444 | |||
D16111.1 | - | 3 | 367 | |||
EU668351.1 | - | 2 | 135 | |||
EU794643.1 | - | 3 | 366 | |||
GQ472218.1 | - | 2 | 135 | |||
HM131449.1 | - | 3 | 358 | |||
S76773.1 | - | 2 | 135 | |||
X75252.1 | - | 3 | 386 | |||
X85033.1 | - | 3 | 279 |