Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030979.2 | NP_112241.2 | 1 | 515 | ||
GenBank mRNA | AF132026.1 | - | 1 | 477 | ||
AK313336.1 | - | 1 | 497 | |||
AL136900.1 | - | 1 | 498 | |||
BC027617.2 | - | 1 | 481 | 4829121 | ||
BC045608.1 | - | 1 | 491 | 5273503 | ||
BG718483.1 | - | 1 | 520 |