Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172477.1 | NP_001165948.1 | 8 | 1090 | ||
NM_001172478.1 | NP_001165949.1 | 7 | 953 | |||
NM_015713.4 | NP_056528.2 | 8 | 1109 | |||
GenBank mRNA | AB036063.1 | - | 8 | 1109 | ||
AB163437.1 | - | 7 | 709 | |||
AB163438.1 | - | 3 | 198 | |||
AB166670.1 | - | 3 | 229 | |||
AB166671.1 | - | 8 | 865 | |||
AK001965.1 | - | 8 | 904 | |||
AK304354.1 | - | 8 | 1034 | |||
AL137348.1 | - | 8 | 911 | |||
BC042468.1 | - | 6 | 667 | |||
BC108261.1 | - | 8 | 921 | 6143754 | ||
BC117496.1 | - | 8 | 946 | 40126047 | ||
BC130628.1 | - | 8 | 946 | 40146659 | ||
BX509409.1 | - | 2 | 113 | |||
CR627376.1 | - | 8 | 1034 |