Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202431.1 | NP_001189360.1 | 4 | 425 | ||
NM_002574.3 | NP_002565.1 | 4 | 642 | |||
NM_181696.2 | NP_859047.1 | 4 | 616 | |||
NM_181697.2 | NP_859048.1 | 4 | 422 | |||
GenBank mRNA | AB451262.1 | - | 3 | 301 | ||
AB451388.1 | - | 3 | 301 | |||
AK131049.1 | - | 4 | 370 | |||
AK311898.1 | - | 4 | 368 | |||
BC007063.1 | - | 4 | 368 | 3961375 | ||
BC021683.1 | - | 4 | 374 | 3681912 | ||
BM740381.1 | - | 4 | 391 | |||
BM762399.1 | - | 4 | 560 | |||
BM787105.1 | - | 4 | 425 | |||
BT019740.1 | - | 3 | 301 | |||
BU564066.1 | - | 4 | 395 | |||
CD638470.1 | - | 4 | 424 | |||
CR407652.1 | - | 3 | 301 | |||
L19184.1 | - | 4 | 368 | |||
X67951.1 | - | 4 | 361 |