Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129888.1 | NP_001123360.1 | 2 | 325 | ||
NM_032687.3 | NP_116076.1 | 2 | 361 | |||
NM_138496.1 | NP_612505.1 | 1 | 186 | |||
XM_006716560.3 | XP_006716623.1 | 2 | 578 | |||
XM_006716561.3 | XP_006716624.1 | 2 | 530 | |||
XM_017013463.1 | XP_016868952.1 | 2 | 586 | |||
XM_017013464.1 | XP_016868953.1 | 2 | 355 | |||
XM_017013465.1 | XP_016868954.1 | 2 | 473 | |||
XM_017013466.1 | XP_016868955.1 | 2 | 355 | |||
GenBank mRNA | AK094514.1 | - | 1 | 186 | ||
BC005073.1 | - | 2 | 361 | 3533499 | ||
BC056412.1 | - | 1 | 639 | |||
BC065484.1 | - | 2 | 325 | 4638402 |