Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016287.3 | NP_057371.2 | 8 | 899 | ||
XM_005245875.4 | XP_005245932.1 | 8 | 1474 | |||
XM_005245876.3 | XP_005245933.1 | 8 | 1007 | |||
XM_005245877.4 | XP_005245934.1 | 8 | 1335 | |||
XM_005245878.4 | XP_005245935.1 | 8 | 1271 | |||
XM_005245879.4 | XP_005245936.1 | 7 | 798 | |||
XM_011541532.1 | XP_011539834.1 | 8 | 991 | |||
XM_011541533.1 | XP_011539835.1 | 8 | 1188 | |||
XM_011541534.2 | XP_011539836.1 | 7 | 808 | |||
XM_017001393.1 | XP_016856882.1 | 8 | 954 | |||
XM_017001394.1 | XP_016856883.1 | 8 | 1319 | |||
GenBank mRNA | AF113534.1 | - | 7 | 832 | ||
AK023129.1 | - | 8 | 899 | |||
AK291252.1 | - | 8 | 877 | |||
AK304065.1 | - | 7 | 775 | |||
AL833978.1 | - | 4 | 715 | |||
BC045660.1 | - | 4 | 1531 | 5271429 | ||
BC046170.1 | - | 8 | 934 | |||
CK000759.1 | - | 5 | 561 |