Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135254.1 | NP_001128726.1 | 7 | 1683 | ||
NM_002584.2 | NP_002575.1 | 7 | 1683 | |||
NM_013945.2 | NP_039236.1 | 7 | 1677 | |||
GenBank mRNA | BC121165.2 | - | 7 | 1159 | 40121582 | |
BC121166.2 | - | 7 | 1159 | 40121584 | ||
DB474828.1 | - | 1 | 100 | |||
DQ322591.1 | - | 7 | 1168 | |||
X96743.1 | - | 7 | 1684 | |||
Z35141.1 | - | 7 | 1187 |