Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016049.3 | NP_057133.2 | 4, 5 | 501 | ||
XM_005267721.4 | XP_005267778.1 | 3, 4 | 1469 | |||
GenBank mRNA | AF151870.1 | - | 4, 5 | 496 | ||
AY368908.1 | - | 4, 5 | 446 | |||
BC002491.2 | - | 4, 5 | 501 | 3051105 |