Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016063.2 | NP_057147.2 | 5 | 690 | ||
GenBank mRNA | AF151888.1 | - | 4 | 447 | ||
AF529363.1 | - | 5 | 486 | |||
BC001671.2 | - | 4 | 522 | |||
BC003357.1 | - | 5 | 529 | |||
BC066332.1 | - | 6 | 504 | 5294646 | ||
BE743302.1 | - | 5 | 603 | |||
BP257644.1 | - | 1 | 77 |