Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002591.3 | NP_002582.3 | 6 | 1001 | ||
XM_011528839.1 | XP_011527141.1 | 4 | 643 | |||
GenBank mRNA | AK290802.1 | - | 6 | 979 | ||
AK303806.1 | - | 4 | 553 | |||
BC023978.1 | - | 6 | 976 | 4042259 | ||
BX648510.1 | - | 1 | 2496 | |||
L05144.1 | - | 6 | 959 |