Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015913.3 | NP_056997.1 | 7 | 1574 | ||
GenBank mRNA | AF131758.1 | - | 7 | 609 | ||
AF543416.1 | - | 7 | 778 | |||
AK056169.1 | - | 7 | 1574 | |||
AK075409.1 | - | 7 | 572 | |||
AY358982.1 | - | 7 | 605 | |||
BC001493.2 | - | 7 | 575 | 3506109 | ||
BC008913.2 | - | 7 | 604 | 3659407 | ||
BC008953.2 | - | 7 | 553 | 2959502 |