Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300848.1 | NP_001287777.1 | 2, 3 | 437 | ||
NM_015950.4 | NP_057034.2 | 2, 3 | 437 | |||
XM_005249161.4 | XP_005249218.1 | 2, 3 | 258 | |||
GenBank mRNA | AF132956.1 | - | 2, 3 | 311 | ||
AK301041.1 | - | 2, 3 | 294 | |||
AK314948.1 | - | 2, 3 | 295 | |||
BC013685.1 | - | 2, 3 | 282 | 3862065 | ||
BC020212.1 | - | 2, 3 | 282 | 4641018 | ||
BM563754.1 | - | 403 | ||||
BM970997.1 | - | 4, 5 | 627 |