Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015956.2 | NP_057040.2 | 8 | 982 | ||
NM_146387.1 | NP_666499.1 | 9 | 846 | |||
NM_146388.1 | NP_666500.1 | 8 | 982 | |||
XM_011528045.2 | XP_011526347.1 | 8 | 977 | |||
GenBank mRNA | AB049635.1 | - | 8 | 697 | ||
AF132962.1 | - | 8 | 943 | |||
AY007152.1 | - | 7 | 635 | |||
BC000756.2 | - | 8 | 936 | 2819730 | ||
BC009858.2 | - | 8 | 723 | 3928448 | ||
BM550378.1 | - | 9 | 844 |