Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015965.6 | NP_057049.5 | 2, 3 | 200 | ||
GenBank mRNA | AF261134.1 | - | 2, 3 | 631 | ||
AF286697.1 | - | 2, 3 | 178 | |||
AK293859.1 | - | 2, 3 | 182 | |||
BC000589.2 | - | 2, 3 | 179 | |||
BC009189.2 | - | 2, 3 | 187 | 3957082 | ||
BG705222.1 | - | 2, 3 | 189 | |||
BM839920.1 | - | 2, 3 | 200 | |||
BP230223.1 | - | 2, 3 | 180 |