Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016094.3 | NP_057178.2 | 5 | 505 | ||
NR_135149.1 | 5 | 585 | ||||
GenBank mRNA | AF059618.1 | - | 5 | 481 | ||
AF125096.1 | - | 5 | 492 | |||
AY542158.1 | - | 5 | 450 | |||
BC001228.1 | - | 5 | 484 | |||
BC022206.1 | - | 5 | 489 | |||
BC046131.1 | - | 5 | 467 | 4443942 | ||
BC093077.1 | - | 5 | 495 | 30372667 | ||
BI792102.1 | - | 5 | 513 | |||
BQ216242.1 | - | 2 | 248 | |||
CN483677.1 | - | 5 | 536 |