Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024940.2 | NP_001020111.1 | 7 | 1548 | ||
NM_016102.3 | NP_057186.1 | 7 | 1584 | |||
XM_006711779.2 | XP_006711842.1 | 7 | 1611 | |||
XM_011544209.2 | XP_011542511.1 | 7 | 1598 | |||
XM_011544210.2 | XP_011542512.1 | 7 | 1700 | |||
XM_011544211.2 | XP_011542513.1 | 7 | 1601 | |||
XM_017001418.1 | XP_016856907.1 | 5 | 2572 | |||
XM_017001419.1 | XP_016856908.1 | 6 | 2116 | |||
GenBank mRNA | AF156271.1 | - | 7 | 1307 | ||
AK055426.1 | - | 6 | 2152 | |||
BC033788.1 | - | 7 | 1348 | 5165609 |