Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039667.2 | NP_001034756.1 | 6 | 1205 | ||
NM_139314.2 | NP_647475.1 | 7 | 1319 | |||
XM_005272484.3 | XP_005272541.1 | 7 | 1371 | |||
XM_005272485.3 | XP_005272542.1 | 6 | 1257 | |||
GenBank mRNA | AB056477.1 | - | 7 | 1293 | ||
AF153606.1 | - | 7 | 1274 | |||
AF169312.1 | - | 7 | 1188 | |||
AF202636.1 | - | 7 | 1296 | |||
AK222489.1 | - | 7 | 1291 | |||
AK222524.1 | - | 7 | 1289 | |||
AK303269.1 | - | 8 | 1220 | |||
AY358275.1 | - | 7 | 1287 | |||
BC023647.2 | - | 7 | 1263 | 5088323 |