Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040138.2 | NP_001035228.1 | 2 | 348 | ||
NM_016951.3 | NP_058647.1 | 2 | 348 | |||
NM_181641.2 | NP_857592.1 | 2 | 348 | |||
GenBank mRNA | AF057306.1 | - | 2 | 326 | ||
AF135380.2 | - | 2 | 342 | |||
AF145216.2 | - | 2 | 342 | |||
AF151058.1 | - | 2 | 345 | |||
AK301240.1 | - | 2 | 339 | |||
AY358999.1 | - | 2 | 325 | |||
BC004380.1 | - | 2 | 331 | 3639550 | ||
DA804889.1 | - | 2 | 329 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204099.1 | NP_001191028.1 | 2 | 348 | ||
GenBank mRNA | FY212695.1 | - | 2 | 232 |