Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016360.3 | NP_057444.2 | 4 | 834 | ||
GenBank mRNA | AK094052.1 | - | 3 | 1670 | ||
AK315375.1 | - | 4 | 834 | |||
AL833861.1 | - | 2 | 247 | |||
BC005049.1 | - | 4 | 822 | 3630410 | ||
BC007744.2 | - | 4 | 646 | 4099917 |