Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135186.1 | NP_001128658.1 | 4 | 955 | ||
NM_016428.2 | NP_057512.1 | 4 | 973 | |||
XM_005257429.2 | XP_005257486.1 | 4 | 618 | |||
XM_011524873.1 | XP_011523175.1 | 4 | 618 | |||
XM_017024721.1 | XP_016880210.1 | 4 | 618 | |||
GenBank mRNA | AB037886.1 | - | 4 | 973 | ||
AL136709.1 | - | 4 | 774 | |||
BC007780.2 | - | 4 | 479 | 4309524 | ||
BI768790.1 | - | 4 | 462 |