Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122646.2 | NP_001116118.2 | 15 | 2075 | ||
NM_001172667.1 | NP_001166138.1 | 3 | 276 | |||
NM_016490.4 | NP_057574.2 | 3 | 394 | |||
XM_011511294.2 | XP_011509596.1 | 14 | 2003 | |||
XM_017004263.1 | XP_016859752.1 | 16 | 2240 | |||
XM_017004264.1 | XP_016859753.1 | 16 | 2201 | |||
XM_017004266.1 | XP_016859755.1 | 6 | 648 | |||
XM_017004267.1 | XP_016859756.1 | 3 | 253 | |||
XM_017004268.1 | XP_016859757.1 | 3 | 279 | |||
GenBank mRNA | AK122705.1 | - | 9 | 954 | ||
AK308698.1 | - | 9 | 957 | |||
BC038583.1 | - | 3 | 355 | |||
BC039488.1 | - | 13 | 1342 | |||
BC051379.1 | - | 3 | 394 | |||
BX103404.1 | - | 3 | 254 |